Cesium sulphate ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10294-54-9
:
Formula:Cs₂O₄S
MDL Number:MFCD00010959
:361.87 g/mol
Merck Index:14,2016
Boiling Pt:1627 °C (1013 hPa)
Melting Pt:1010 °C
Density:4.243
Storage Temperature:Ambient
EINECS:233-662-6
Related Information
Frequently Bought Together