Potassium dihydrogen phosphate
Catalog # SERA26870.01
Supplier: Serva
CAS Number:
Specifications
- Verpakking:Glass bottle for solids
- VPE:500 g
- Seq:3645
Specifications
About this item
:
:136.09 g/mol
Merck Index:13,07744
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:253 °C
Density:2,3 g/cm³ (20 °C)
Storage Temperature:Kamertemperatuur
EINECS:231-913-4
REACH:01-2119490224-41
CAS Number:7778-77-0
Formula:KH₂PO₄
MDL Number:MFCD00011401