Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Potassium hexacyanoferrate(III)

Marketsource
Supplier:   Roth Carl
Potassium hexacyanoferrate(III)
Supplier:  Roth Carl
Marketsource
Certificates
Potassium hexacyanoferrate(III)

2 Options Available Below

Product Details & Documents
Synonyms:  Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide
  • CAS Number:13746-66-2
  • Formula:K₃Fe(CN)₆
  • MDL Number:MFCD00011392
  • Molecular Weight:329.25 g/mol
  • Merck Index:13,07712
  • Density:1.85 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:237-323-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 139,00 €
    139,00 €
    1 * 500 g
       139,00 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 259,00 €
    259,00 €
    1 * 1 kg
       259,00 €

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.