Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Sodium hydrogen carbonate

Sodium hydrogen carbonate

Marketsource
Supplier:   Roth Carl
Sodium hydrogen carbonate
Supplier:  Roth Carl
Marketsource
Certificates
Sodium hydrogen carbonate
Sodium hydrogen carbonate

4 Options Available Below

Product Details & Documents
Synonyms:  Sodium bicarbonate
  • CAS Number:144-55-8
  • Formula:NaHCO₃
  • MDL Number:MFCD00003528
  • Molecular Weight:84.01 g/mol
  • Merck Index:13,08655
  • Melting Pt:270 °C
  • Density:2.159 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-633-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

    Specifications:
    Marketsource
    Marketsource
  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg

    Specifications:
    Marketsource
    Marketsource
  • 25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg

    Specifications:
    Marketsource
    Marketsource