2-Furaldehyde
Catalog # MAYBCC02004.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:96.09 g/mol
Merck Index:13,04325
Boiling Pt:162 °C (1013 hPa)
Melting Pt:–39 °C
Density:1.15 g/cm³ (20 °C)
Flash Pt:60 °C
Storage Temperature:Ambient
EINECS:202-627-7
UN:1199
ADR:6.1,II
:GHS06
:GHS08
CAS Number:98-01-1
Formula:C₅H₄O₂
MDL Number:MFCD00003229