2-Furaldehyde ≥98% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-01-1
:
Formula:C₅H₄O₂
MDL Number:MFCD00003229
:96.09 g/mol
Merck Index:14,4304
Boiling Pt:162 °C
Melting Pt:–38 °C
Density:1.159
Flash Pt:60 °C (140 °F)
EINECS:202-627-7
UN:1199
ADR:6.1,II
:GHS06
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together