1,5-Diphenylcarbazide ACS, Reag. Ph. Eur. analytical reagent, Supelco®
: Merck
:140-22-7
:
:C₁₃H₁₄N₄O
:MFCD00003013
:242.28 g/mol
:13,03353
:454 °C (1013 hPa)
:170…172 °C
:1.31 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:205-403-7
:GHS07
:Ambient
- Solubility:(20 °C) slightly soluble
- Melting Point:173 - 176 °C
- Molar Mass:242.28 g/mol
- Bulk Density:420 kg/m3
Frequently Bought Together