Order Entry
Netherlands
ContactUsLinkComponent
2-Phenylaniline ≥99.0% (by GC, titration analysis) for sugar determination
Irritant
Health
2-Phenylaniline ≥99.0% (by GC, titration analysis) for sugar determination
Supplier:  TCI
Certificates
undefined
Irritant
Health
2-Phenylaniline ≥99.0% (by GC, titration analysis) for sugar determination
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Melting point:
    48.0 to 51.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together