Supelco® Organic Reference Standard, PCB No 209
Catalog # 31092-20MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:20 mg
- Standards:Analytical standard
- CAS No.:2051-24-3
- Description:PCB No 209
- Storage conditions:2...30 °C
- Hazard class description:H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055243
- M-Clarity quality level:MQ100
- Synonym(s):Decachlorobiphenyl; 2,2′,3,3′,4,4′,5,5′,6,6′-PCB
- Molecular weight:498,66
- Flash point:>100,0 °C
Specifications
About this item
- GHS signal: GHS08, GHS09
Caution: P260 - P273 - P314 - P391 - P501