Order Entry
Netherlands
ContactUsLinkComponent
3,4',5-Trichlorobiphenyl (PCB No. 39)
3,4',5-Trichlorobiphenyl (PCB No. 39)
Catalog # PROOCIL-PCB-39-C
CAS Number:  
3,4',5-Trichlorobiphenyl (PCB No. 39)
Catalog # PROOCIL-PCB-39-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :257.55 g/mol
CAS Number:38444-88-1
Formula:C₁₂H₇Cl₃