Order Entry
Netherlands
ContactUsLinkComponent
2,2',3,4,6,6'-Hexachlorobiphenyl (PCB No. 145)
2,2',3,4,6,6'-Hexachlorobiphenyl (PCB No. 145)
Catalog # PROOCIL-PCB-145-C
CAS Number:  
2,2',3,4,6,6'-Hexachlorobiphenyl (PCB No. 145)
Catalog # PROOCIL-PCB-145-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :360.88 g/mol
UN:3432
ADR:9,II
CAS Number:74472-40-5
Formula:C₁₂H₄Cl₆