Order Entry
Netherlands
ContactUsLinkComponent
2,2',3,4',6-Pentachlorobiphenyl (PCB No. 91)
2,2',3,4',6-Pentachlorobiphenyl (PCB No. 91)
Catalog # PROOCIL-PCB-91-C
CAS Number:  
2,2',3,4',6-Pentachlorobiphenyl (PCB No. 91)
Catalog # PROOCIL-PCB-91-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :326.44 g/mol
CAS Number:68194-05-8
Formula:C₁₂H₅Cl₅