Supelco® Organic Reference Standard, PCB No 52
35599-10MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:35693-99-3
- Description:PCB No 52
- Storage conditions:2...30 °C
- Hazard class description:H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055544
- M-Clarity quality level:MQ100
- Synonym(s):2,2′,5,5′-Tetrachlorobiphenyl; 2,2′,5,5′-PCB
- Molecular weight:291,99
- Flash point:Not applicable
- GHS signal: GHS08, GHS09
: P260 - P273 - P314 - P391 - P501