Supelco® Organic Reference Standard, PCB No 12
35593-100MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:2974-92-7
- Description:PCB No 12
- Storage conditions:2...30 °C
- Hazard class description:H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055250
- M-Clarity quality level:MQ100
- Synonym(s):3,4-Dichlorobiphenyl; 3,4-PCB
- Molecular weight:223,10
- Flash point:Not applicable
: P260 - P273 - P314 - P391 - P501