2,2',3,3',6,6'-Hexachlorobiphenyl (PCB No. 136)
Catalog # EHERC20013600
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:20 mg
- Seq:2305
Specifications
About this item
:
:360.88 g/mol
CAS Number:38411-22-2
Formula:C₁₂H₄Cl₆