Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,3',4,4',5'-Hexachlorobiphenyl (PCB No. 157)
2,3,3',4,4',5'-Hexachlorobiphenyl (PCB No. 157)
2,3,3',4,4',5'-Hexachlorobiphenyl (PCB No. 157)
PROOCIL-PCB-157-C
:
LGC Standards PROMOCHEM
:
2,3,3',4,4',5'-Hexachlorobiphenyl (PCB No. 157)
PROOCIL-PCB-157-C
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle for solids
Pk:
5 mg
Seq:
2210
:
:
9,II
:
360.88 g/mol
:
3432
:
69782-90-7
:
C₁₂H₄Cl₆