4,4'-Di-tert-butylbiphenyl 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1625-91-8
:
Formula:C₂₀H₂₆
MDL Number:MFCD00008834
Boiling Pt:190…192 °C (13 mmHg)
Melting Pt:126…129 °C
Density:0,9691 g/cm³ (20 °C)
:266.43 g/mol
Storage Temperature:Ambient
EINECS:216-615-4
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:126 to 129 °C
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together