Order Entry
Netherlands
ContactUsLinkComponent
2-(Hydroxymethyl)benzyl alcohol for synthesis, Sigma-Aldrich®
2-(Hydroxymethyl)benzyl alcohol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
2-(Hydroxymethyl)benzyl alcohol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Melting Point:
    62 - 64 °C
  • Molar Mass:
    138.16 g/mol
  • Synonyms:
    1,2-Bishydroxymethylbenzene
  • Hill Formula:
    C8H10O2

Specification Test Results

Related Information

Frequently Bought Together