4-Fluoro-o-xylene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-Fluoro-3,4-dimethylbenzene, 4-Fluoro-1,2-dimethylbenzene
CAS Number:452-64-2
:
Formula:C₈H₉F
MDL Number:MFCD00039217
Boiling Pt:148…149 °C
Density:1.000
Molecular Weight:124.16 g/mol
Flash Pt:40 °C (104 °F)
Storage Temperature:Ambient
UN:1993
ADR:3 III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together