Order Entry
Netherlands
ContactUsLinkComponent
 
4'-Methylacetophenone 95%
 
undefined
4'-Methylacetophenone 95%

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5300 to 1.5350 (20°C, 589 nm)
  • GC:
    ≥94 %

 

 

Frequently Bought Together