5-Bromo-m-xylene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-Bromo-3,5-dimethylbenzene
CAS Number:556-96-7
:
Formula:C₈H₉Br
MDL Number:MFCD00000087
Boiling Pt:204 °C
Density:1.361
Molecular Weight:185.06 g/mol
Flash Pt:87 °C (188 °F)
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together