1,2-Diphenylethane 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:103-29-7
:
Formula:(C₆H₅)CH₂CH₂(C₆H₅)
MDL Number:MFCD00004796
:182.27 g/mol
Merck Index:12,01245
Boiling Pt:284 °C (760 mmHg)
Melting Pt:49…53 °C
Density:1.01 g/cm³
Flash Pt:129 °C
Storage Temperature:Ambient
EINECS:203-096-4
Specification Test Results
- Appearance:White to light yellow Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:49 to 53 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together