

p-Toluenesulphonyl hydrazide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1576-35-8
:
Formula:C₇H₁₀N₂O₂S
MDL Number:MFCD00007588
:186.23 g/mol
Boiling Pt:332 °C (1013 hPa)
Melting Pt:109…113 °C
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-407-3
UN:3226
ADR:4.1,
:GHS06
:GHS02
Specification Test Results
- Appearance:White to off-white to light grey Powder
- Infrared spectrum:Conforms
- Melting point:105 to 113 °C (Dec)
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together