Toluene-4-sulphonamide
Catalog # EHERC17594700
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
Boiling Pt:322 °C (1013 hPa)
Melting Pt:136…138 °C
:171.22 g/mol
Storage Temperature:Ambient
EINECS:200-741-1
CAS Number:70-55-3
Formula:C₇H₉NO₂S
MDL Number:MFCD00011692