Order Entry
Netherlands
ContactUsLinkComponent
2-Nitro-p-toluidine 99%
Toxic
Health
Environment
2-Nitro-p-toluidine 99%
Certificates
undefined
Toxic
Health
Environment
2-Nitro-p-toluidine 99%

Specification Test Results

  • Appearance:
    Orange to orange-brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    112 to 116 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together