4-Nitro-o-toluidine
Catalogus # MOLE31882578-500G
Supplier: Molekula
CAS Number:
Specifications
- Verpakking:Glass bottle for solids
- VPE:500 g
- Seq:3645
Specifications
About this item
:
:152.15 g/mol
Melting Pt:135 °C
Density:1,366 g/cm³ (15 °C)
Storage Temperature:Kamertemperatuur
EINECS:202-762-1
UN:2660
ADR:6.1,III
:GHS06
:GHS08
:GHS09
CAS Number:99-52-5
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007734