m-Tolyl isothiocyanate
Marketsource
Supplier: Thermo Scientific
Jump to:
About this item
- CAS Number:621-30-7
- Formula:C₈H₇NS
- MDL Number:MFCD00004807
- Molecular Weight:149.22 g/mol
- Boiling Pt:243 °C (1013 hPa)
- Melting Pt:–20 °C
- Density:1,103 g/cm³ (20 °C)
- EINECS:210-677-6
- UN:2922
- ADR:8,II
3 Options Available Below
Product Details & Documents
- CAS Number:621-30-7
- Formula:C₈H₇NS
- MDL Number:MFCD00004807
- Molecular Weight:149.22 g/mol
- Boiling Pt:243 °C (1013 hPa)
- Melting Pt:–20 °C
- Density:1,103 g/cm³ (20 °C)
- EINECS:210-677-6
- UN:2922
- ADR:8,II
Product Family Options
Product Information
- VerpakkingVPEAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



