

p-Tolyl isocyanate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:622-58-2
:
Formula:C₈H₇NO
MDL Number:MFCD00002029
:133.15 g/mol
Boiling Pt:70…72 °C (10 mmHg)
Density:1.059
Flash Pt:66 °C (150 °F)
EINECS:210-743-4
UN:2206
ADR:6.1,III
:GHS08
:GHS07
Related Information
Frequently Bought Together