Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
Netherlands
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
ContactUsLinkComponent
Home
α,α,α-Trifluoro-p-cresol
α,α,α-Trifluoro-p-cresol
α,α,α-Trifluoro-p-cresol
α,α,α-Trifluoro-p-cresol
PROOMM0256.05
:
LGC Standards PROMOCHEM
:
α,α,α-Trifluoro-p-cresol
PROOMM0256.05
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle for solids
Pk:
100 mg
Seq:
2440
:
:
162.11 g/mol
:
185 °C (1013 hPa)
:
45…47 °C
:
1,3 g/cm³ (25 °C)
:
Refrigerator
:
206-945-7
:
GHS07
:
402-45-9
:
C₇H₅F₃O
:
MFCD00002363