6-Fluoro-m-toluidine
Catalog # MOLE31695268-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:125.15 g/mol
Boiling Pt:88…90 °C (17 mmHg)
Density:1.109 g/cm³ (20 °C)
UN:2810
ADR:6.1,III
:GHS07
CAS Number:452-84-6
Formula:C₇H₈FN
MDL Number:MFCD00007654