4-(Trifluoromethyl)phenyl isothiocyanate
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:1645-65-4
:
Formula:C₈H₄F₃NS
MDL Number:MFCD00039645
:203.19 g/mol
Boiling Pt:205…207 °C (1013 hPa)
Melting Pt:29…31 °C
UN:2923
ADR:8,III
Frequently Bought Together