

4-Chloro-2-(trifluoromethyl)phenyl isocyanate 97%
Catalog # L09850.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:16588-69-5
:
Formula:C₈H₃ClF₃NO
:221.57 g/mol
Boiling Pt:114…115 °C (10 mmHg)
Density:1.467
Flash Pt:94 °C (201 °F)
UN:2206
ADR:6.1,III
:GHS07
:GHS08
MDL Number:MFCD00013873