4-Chloro-2-(trifluoromethyl)phenyl isocyanate
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:16588-69-5
:
Formula:C₈H₃ClF₃NO
MDL Number:MFCD00013873
:221.57 g/mol
Boiling Pt:85…90 °C (13 mmHg)
Density:1.4666 g/cm³ (20 °C)
UN:2206
ADR:6.1,III
Frequently Bought Together