2-Phenoxyethanol
MOLEM30650496
: Molekula
:
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
:
:138.17 g/mol
:12,07410
:245 °C (1013 hPa)
:13 °C
:1.1094 g/cm³ (20 °C)
:121 °C (closed cup)
:Ambient
:204-589-7
:GHS07
:122-99-6
:C₆H₅OCH₂CH₂OH
:MFCD00002857