


Diphenylamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:122-39-4
:
Formula:(C₆H₅)₂NH
MDL Number:MFCD00003014
:169.23 g/mol
Merck Index:13,03349
Boiling Pt:302 °C (1013 hPa)
Melting Pt:53…54 °C
Density:1.165 g/cm³ (20 °C)
EINECS:204-539-4
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
Flash Pt:153 °C (closed cup)
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.05 g/l (25 °C)
- Melting Point:53 - 54 °C
- Molar Mass:169.23 g/mol
- Bulk Density:610 kg/m3
- Boiling Point:302 °C (1013 hPa)
- Vapor Pressure:0.0003 hPa (20 °C)
- Flash Point:153 °C DIN 51758
- Density:1.16 g/cm3 (20 °C)
- Ignition Point:630 °C DIN 51794
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.20528.1000_TDS_EN ProductPage_820528
Product specification
msds_820528
SDS
Frequently Bought Together