Diphenylamine, Supelco®
Glass bottle for solids, 250 mg
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: N-Phenylaniline, N-Phenylbenzeneamine
- CAS Number:122-39-4
- Formula:(C₆H₅)₂NH
- MDL Number:MFCD00003014
- Molecular Weight:169.23 g/mol
- Merck Index:13,03349
- Boiling Pt:302 °C (1013 hPa)
- Melting Pt:53…54 °C
- Density:1.165 g/cm³ (20 °C)
- Flash Pt:153 °C (closed cup)
- Storage Temperature:Ambient
- UN:3077
- ADR:9,III
- :M-Clarity™ quality level = MQ100
- EINECS:204-539-4
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



