
4-Fluorobenzenesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:349-88-2
:
Formula:C₆H₄ClFO₂S
MDL Number:MFCD00007438
:194.61 g/mol
Boiling Pt:86…87 °C (1 mmHg)
Melting Pt:35…38 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:206-493-0
UN:3261
ADR:8,II
:GHS05
Related Information
Frequently Bought Together