Order Entry
Netherlands
ContactUsLinkComponent
Potassium hydrogen phthalate ≥99.5% for acid-base titration, Supelco®
Potassium hydrogen phthalate ≥99.5% for acid-base titration, Supelco®
Catalog # SUPL60357-50G
Supplier:  Merck
CAS Number:  
Potassium hydrogen phthalate ≥99.5% for acid-base titration, Supelco®
Catalog # SUPL60357-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :204.22 g/mol
Merck Index:13,07694
Melting Pt:295…300 °C
Density:1,6362 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-889-4
CAS Number:877-24-7
Formula:HOOCC₆H₄COOK
MDL Number:MFCD00013070