Order Entry
Netherlands
ContactUsLinkComponent
Terephthalic acid for synthesis, Sigma-Aldrich®
Terephthalic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Terephthalic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    15 mg/l (20 °C) (experimental)
  • Melting Point:
    402 °C (sublimed)
  • Molar Mass:
    166.13 g/mol
  • Density:
    1.51 g/cm3 (20 °C)
  • Ignition Point:
    496 °C

Specification Test Results

Related Information

Frequently Bought Together