Order Entry
Netherlands
ContactUsLinkComponent
Dibutyl phthalate ≥99%, TECHNICAL
Dibutyl phthalate ≥99%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Dibutyl phthalate ≥99%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Acidity:
    Max. 0.003 meq/g
  • Ignition residue (SO4):
    Max. 0.1 %
  • Water:
    Max. 0.2 %

Specification Test Results

Frequently Bought Together