
2,5-Dichlorophenyl isocyanate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5392-82-5
:
Formula:C₇H₃Cl₂NO
MDL Number:MFCD00002002
:188.01 g/mol
Boiling Pt:242 °C (1013 hPa)
Melting Pt:19 °C
Storage Temperature:Refrigerator
EINECS:226-396-7
UN:2250
ADR:6.1,II
:GHS06
Specification Test Results
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (GC):min. 97.0 %
- Freezing point:22.0 to 26.0 deg-C
Specification Test Results
Frequently Bought Together