3,5-Dichlorophenyl isocyanate for synthesis
Supplier: Merck
Certificates
About this item
CAS Number:34893-92-0
:
Formula:C₇H₃Cl₂NO
MDL Number:MFCD00013859
:188.01 g/mol
Boiling Pt:264 °C (1013 hPa)
Melting Pt:32…34 °C
Density:1.38 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:252-276-9
UN:2250
ADR:6.1,II
Frequently Bought Together