
3,4-Dichlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
TCIAD0346-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
:
:209.46 g/mol
:242 °C (1013 hPa)
:34 °C
:Ambient
:221-177-2
:3261
:8,III
:GHS05
:3024-72-4
:C₇H₃Cl₃O
:MFCD00000672