Order Entry
Netherlands
ContactUsLinkComponent
 
 
2,5-Dichlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
2,5-Dichlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
 :  TCI
 

 

  • Appearance:
    White or Colorless to Light orange to Yellow powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Solubility in Toluene:
    almost transparency

 

 

Frequently Bought Together