
2,3-Dichlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2905-60-4
:
Formula:C₇H₃Cl₃O
MDL Number:MFCD00035937
:209.46 g/mol
Boiling Pt:140°C
Melting Pt:28…28.5 °C
EINECS:220-811-5
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in Toluene:very faint turbidity
Specification Test Results
Frequently Bought Together