Order Entry
Netherlands
ContactUsLinkComponent
3-Chlorophenylboronic acid
3-Chlorophenylboronic acid
Catalog # MOLE66647138-5G
Supplier:  Molekula
CAS Number:  
3-Chlorophenylboronic acid
Catalog # MOLE66647138-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :156.38 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:63503-60-6
Formula:C₆H₆BClO₂
MDL Number:MFCD00161354