


2-Bromophenyl isocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1592-00-3
:
Formula:C₇H₄BrNO
MDL Number:MFCD00001995
:198.02 g/mol
Boiling Pt:91…92 °C (5 mmHg)
Density:1,607
Flash Pt:107 °C (224 °F)
UN:2206
ADR:6.1,II
:GHS06
:GHS08
:GHS07
Related Information
Frequently Bought Together