

4-Bromo-2,5-difluoroaniline ≥98%
Catalog # B24457.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:208.01 g/mol
Melting Pt:74…75 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
:GHS07
CAS Number:112279-60-4
Formula:C₆H₄BrF₂N
MDL Number:MFCD01861125