Methyl-2-bromobenzoate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:610-94-6
:
Formula:C₈H₇BrO₂
MDL Number:MFCD00016328
Boiling Pt:252 °C
Density:1.532
Flash Pt:>110 °C (230 °F)
:215.05 g/mol
Storage Temperature:Ambient
EINECS:210-241-5
Related Information
Frequently Bought Together