Morpholine, Supelco®
Glass bottle, 10 mL
Catalog # 90179-10ML-F
Supplier: Merck
Jump to:
Product Details & Documents
- CAS Number:110-91-8
- Formula:C₄H₉NO
- MDL Number:MFCD00005972
- Molecular Weight:87.12 g/mol
- Merck Index:12,06362
- Boiling Pt:128.9 °C (1013 hPa)
- Melting Pt:–5 °C
- Density:1 g/cm³ (20 °C)
- Flash Pt:38 °C
- Storage Temperature:Ambient
- UN:2054
- ADR:8,I
- :M-Clarity™ quality level = MQ100
- EINECS:203-815-1
Specifications
Specifications
- Pack type:Glass bottle
- Pk:10 mL



